Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pomalidomide 4'-PEG1-azide
GREENER_CHOICESDP

Catalog No. 743425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-342-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 74-342-5 Supplier Tocris Bioscience™ Supplier No. 7434/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cereblon ligand with PEG linker and terminal azide for onward chemistry

Functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material 4-[[2-(2-Azidoethoxy)ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
CAS 2271036-44-1
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C17H18N6O5
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.