Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pomalidomide
GREENER_CHOICE

Catalog No. 630210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-021-0 10 mg
63-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-021-0 Supplier Tocris Bioscience™ Supplier No. 6302/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cereblon binder; also TNF-α inhibitor and antiangiogenic

Pomalidomide is a potent TNF-α inhibitor (IC50 = 13 nM). Also potently inhibits IL-2 (EC50 = 8 nM). Thalidomide derivative. Binds cereblon and inhibits its ubiquitination. Exhibits antiproliferative effects in a Namalwa lymphoma cell line. Antiangiogenic. Promotes degradation of transcription factor SALL4.

Specifications

Chemical Name or Material 4-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
CAS 19171-19-8
Assay Percent Range >98%
Quantity 10 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C13H11N3O4
Formula Weight Observed MW: 273.24
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.