Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMC Antibody (OTI2B2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273525FR
Description
POMC Monoclonal specifically detects POMC in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POMC | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
POMC | |
Full length recombinant protein of human POMC (NP_001030333) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2B2 | |
Western Blot, Immunohistochemistry | |
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
Mouse | |
Affinity Purified | |
Neuroscience, Nutrient Sensing in the Brain | |
5443.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction