Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POMC Rabbit anti-Human, Unconjugated, Clone: CLIP/2859R, Abnova™

Catalog No. 89339584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-584 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-584 Supplier Abnova Corporation Supplier No. RAB00605
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

POMC Monoclonal antibody specifically detects POMC in Human samples. It is validated for ELISA.

Specifications

Antigen POMC
Applications ELISA
Classification Monoclonal
Clone CLIP/2859R
Conjugate Unconjugated
Description proopiomelanocortin
Gene POMC
Gene Alias ACTH, CLIP, LPH, MSH, NPP, POC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 25-39 of human POMC
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5443
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.