Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMGNT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560735
Description
Immunogen sequence: LFLNMVAPGR VLICTVKDEG SFHLKDTAKA LLRSLGSQAG PALGWRDTWA FVGRKGGPVF GEKHSKSPAL SSWGDPVLLK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The product of the POMGNT1 gene, protein O-mannose beta-1,2-N-acetylglucosaminyltransferase, participates in O-mannosyl glycan synthesis. POMGnT1 is an N(in)/C(out) (type II) membrane protein localized in the medial-Golgi that initiates the conversion of high mannose N-glycans to complex N-glycans. Specifically, POMGnT1 is a glycosylation enzyme that participates in the synthesis of O-mannosyl glycan, a laminin-binding ligand of alpha-dystroglycan that is rarely synthesized in mammals. Mutations in the POMGNT1 gene cause muscle-eye-brain disease (MEB), an autosomal recessive disorder characterized by congenital muscular dystrophy, ocular abnormalities and lissencephaly.
Specifications
POMGNT1 | |
Polyclonal | |
Unconjugated | |
POMGNT1 | |
0610016I07Rik; 4930467B06Rik; GnT I.2; GNTI.2; gnT-I.2; LGMD2O; MDDGA3; MDDGB3; MDDGC3; MEB; MGAT1.2; N-acetylglucosaminyltransferase I.2; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; OMGnT1; POMGNT1; protein O-linked mannose beta 1,2-N-acetylglucosaminyltransferase; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-); protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; LOW QUALITY PROTEIN: protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2; UNQ746/PRO1475 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WZA1 | |
POMGNT1 | |
Recombinant protein corresponding to Human POMGNT1. Recombinant protein control fragment (Product #RP-94043). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction