Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMGNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569556 Supplier Invitrogen™ Supplier No. PA569556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.

The product of the POMGNT1 gene, protein O-mannose beta-1,2-N-acetylglucosaminyltransferase, participates in O-mannosyl glycan synthesis. POMGnT1 is an N(in)/C(out) (type II) membrane protein localized in the medial-Golgi that initiates the conversion of high mannose N-glycans to complex N-glycans. Specifically, POMGnT1 is a glycosylation enzyme that participates in the synthesis of O-mannosyl glycan, a laminin-binding ligand of alpha-dystroglycan that is rarely synthesized in mammals. Mutations in the POMGNT1 gene cause muscle-eye-brain disease (MEB), an autosomal recessive disorder characterized by congenital muscular dystrophy, ocular abnormalities and lissencephaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMGNT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POMGNT1
Gene Accession No. Q91X88
Gene Alias 0610016I07Rik; 4930467B06Rik; GnT I.2; GNTI.2; gnT-I.2; LGMD2O; MDDGA3; MDDGB3; MDDGC3; MEB; MGAT1.2; N-acetylglucosaminyltransferase I.2; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; OMGnT1; POMGNT1; protein O-linked mannose beta 1,2-N-acetylglucosaminyltransferase; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-); protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; LOW QUALITY PROTEIN: protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2; UNQ746/PRO1475
Gene Symbols POMGNT1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse POMGNT1 (aa 47-96).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68273
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.