Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMGNT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576448
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The product of the POMGNT1 gene, protein O-mannose beta-1,2-N-acetylglucosaminyltransferase, participates in O-mannosyl glycan synthesis. POMGnT1 is an N(in)/C(out) (type II) membrane protein localized in the medial-Golgi that initiates the conversion of high mannose N-glycans to complex N-glycans. Specifically, POMGnT1 is a glycosylation enzyme that participates in the synthesis of O-mannosyl glycan, a laminin-binding ligand of alpha-dystroglycan that is rarely synthesized in mammals. Mutations in the POMGNT1 gene cause muscle-eye-brain disease (MEB), an autosomal recessive disorder characterized by congenital muscular dystrophy, ocular abnormalities and lissencephaly.
Specifications
POMGNT1 | |
Polyclonal | |
Unconjugated | |
POMGNT1 | |
0610016I07Rik; 4930467B06Rik; GnT I.2; GNTI.2; gnT-I.2; LGMD2O; MDDGA3; MDDGB3; MDDGC3; MEB; MGAT1.2; N-acetylglucosaminyltransferase I.2; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; OMGnT1; POMGNT1; protein O-linked mannose beta 1,2-N-acetylglucosaminyltransferase; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-); protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; LOW QUALITY PROTEIN: protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2; UNQ746/PRO1475 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362567, 55624, 68273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5XIN7, Q8WZA1, Q91X88 | |
POMGNT1 | |
Recombinant fusion protein of human POMGNT1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction