Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMGNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116590 Supplier Invitrogen™ Supplier No. PA5116590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, 293T, BxPC-3, A-549, Mouse ovary, Mouse brain, Mouse pancreas, Rat brain Immunogen sequence: NPDNLMHVFH DDLLPLFYTL RQFPGLAHEA RLFFMEGWGE GAHFDLYKLL SPKQPLLRAQ LKTLGRLLCF SHAFVGLSKI TTWYQYGFVQ PQGPKANILV SGNEIRQFAR FMTEKLNVSH TGVPLGEEYI LVFSRTQNRL ILNEAELLLA LAQEFQMKTV TVSLEDHTFA DVVRLVSNAS M.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMGNT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene POMGNT2
Gene Accession No. Q5NDF0, Q8BW41, Q8NAT1
Gene Alias ago61; C22H3orf39; C23H3orf39; C3orf39; C85492; Eogtl; Extracellular O-linked N-acetylglucosamine transferase-like; glycosyltransferase; glycosyltransferase ago61; glycosyltransferase-like domain containing 2; glycosyltransferase-like domain-containing protein 2; GTDC2; MDDGA8; Pomgnt2; protein O-linked mannose beta 1,4-N-acetylglucosaminyltransferase 2; protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-); protein O-linked-mannose beta-1,4-N-acetylglucosaminyltransferase 2; RGD1304827
Gene Symbols POMGNT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-340 of human POMGNT2 (NP_1161952).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215494, 316091, 84892
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.