Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMK Monoclonal Antibody (OTI1B1)
GREENER_CHOICE

Catalog No. PIMA526679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526679 Supplier Invitrogen™ Supplier No. MA526679
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POMK Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein that may be involved in the presentation of the laminin-binding O-linked carbohydrate chain of alpha-dystroglycan (a-DG), which forms transmembrane linkages between the extracellular matrix and the exoskeleton. Some pathogens use this O-linked carbohydrate unit for host entry. Loss of function compound heterozygous mutations in this gene were found in a human patient affected by the Walker-Warburg syndrome (WWS) phenotype. Mice lacking this gene contain misplaced neurons (heterotopia) in some regions of the brain, possibly from defects in neuronal migration. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POMK
Gene Accession No. Q9H5K3
Gene Alias 4930444A02Rik; MDDGA12; MDDGC12; Pomk; probable inactive protein kinase-like protein SgK196; protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; protein O-mannose kinase; protein-O-mannose kinase; RGD1310810; SGK196; sugen kinase 196
Gene Symbols POMK
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 141-350 of SGK196 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84197
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.