Learn More
Invitrogen™ POMK Monoclonal Antibody (OTI2B10)

Description
POMK Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | POMK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2B10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | POMK |
| Gene Accession No. | Q9H5K3 |
| Gene Alias | 4930444A02Rik; MDDGA12; MDDGC12; Pomk; probable inactive protein kinase-like protein SgK196; protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; protein O-mannose kinase; protein-O-mannose kinase; RGD1310810; SGK196; sugen kinase 196 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.