Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553037
Description
Immunogen sequence: TSNFSILAND LDALPLVNHS SGMLVKCGHR ELHGDFVAPE QLWPYGEDVP FHDDLMPSYD EKIDIWKIPD ISSFLLGHIE GSDMVRFHLF DIHKACKSQT PSERPTAQDV LETYQKVLDT L Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
This gene encodes a protein that may be involved in the presentation of the laminin-binding O-linked carbohydrate chain of alpha-dystroglycan (a-DG), which forms transmembrane linkages between the extracellular matrix and the exoskeleton. Some pathogens use this O-linked carbohydrate unit for host entry. Loss of function compound heterozygous mutations in this gene were found in a human patient affected by the Walker-Warburg syndrome (WWS) phenotype. Mice lacking this gene contain misplaced neurons (heterotopia) in some regions of the brain, possibly from defects in neuronal migration. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| POMK | |
| Polyclonal | |
| Unconjugated | |
| POMK | |
| 4930444A02Rik; MDDGA12; MDDGC12; Pomk; probable inactive protein kinase-like protein SgK196; protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; protein O-mannose kinase; protein-O-mannose kinase; RGD1310810; SGK196; sugen kinase 196 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84197 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H5K3 | |
| POMK | |
| Recombinant protein corresponding to Human POMK. Recombinant protein control fragment (Product #RP-90559). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction