Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMK/SGK196 Antibody (OTI2B10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272275FR
Description
POMK/SGK196 Monoclonal specifically detects POMK/SGK196 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POMK/SGK196 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
POMK | |
Recombinant protein fragment corresponding to amino acids 141-350 of human FLJ23356 (NP_115613) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2B10 | |
Western Blot, Immunohistochemistry | |
FLJ23356, MGC126597, protein kinase-like protein SgK196, Sugen kinase 196 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
84197.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction