Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMP Polyclonal Antibody

Catalog No. PIPA5121673
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121673 20 μL
PIPA5119365 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121673 Supplier Invitrogen™ Supplier No. PA5121673
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POMP Polyclonal Antibody for Western Blot, IHC (P)

Molecular chaperone essential for the assembly of standard proteasomes and immunoproteasomes. Degraded after completion of proteasome maturation. Mediates the association of 20S preproteasome with the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene POMP
Gene Accession No. Q9Y244
Gene Alias 2510048O06Rik; C13orf12; HSPC014; HSPC036; hUMP1; mUMP1; PNAS-110; POMP; Proteasome maturation protein; proteassemblin; Protein UMP1 homolog; RGD1305831; UMP1; Voltage-gated K channel beta subunit 4.1; voltage-gated potassium channel beta subunit 4.1
Gene Symbols POMP
Host Species Rabbit
Immunogen Recombinant protein Proteasome Maturation Protein. The antigen corresponds to amino acid range 1-141 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102163533, 51371
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.