Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555478
Description
Immunogen sequence: SELKDSIPVT ELSASGPFES HDLLRKGFSC VKNELLPSHP LELSEKNFQL NQDKMNFSTL RNIQGLFAPL KLQMEFKAVQ QVQRLPFLSS SNLSLDVLRG NDETIGFE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
Molecular chaperone essential for the assembly of standard proteasomes and immunoproteasomes. Degraded after completion of proteasome maturation. Mediates the association of 20S preproteasome with the endoplasmic reticulum.
Specifications
| POMP | |
| Polyclonal | |
| Unconjugated | |
| POMP | |
| 2510048O06Rik; C13orf12; HSPC014; HSPC036; hUMP1; mUMP1; PNAS-110; POMP; Proteasome maturation protein; proteassemblin; Protein UMP1 homolog; RGD1305831; UMP1; Voltage-gated K channel beta subunit 4.1; voltage-gated potassium channel beta subunit 4.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y244 | |
| POMP | |
| Recombinant protein corresponding to Human POMP. Recombinant protein control fragment (Product #RP-91587). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction