Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117151 Supplier Invitrogen™ Supplier No. PA5117151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, HepG2, Jurkat Immunogen sequence: MNARGLGSEL KDSIPVTELS ASGPFESHDL LRKGFSCVKN ELLPSHPLEL SEKNFQLNQD KMNFSTLRNI QGLFAPLKLQ MEFKAVQQVQ RLPFLSSSNL SLDVLRGNDE TIGFEDILND PSQSEVMGEP HLMVEYKLGL L.

Molecular chaperone essential for the assembly of standard proteasomes and immunoproteasomes. Degraded after completion of proteasome maturation. Mediates the association of 20S preproteasome with the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene POMP
Gene Accession No. Q9Y244
Gene Alias 2510048O06Rik; C13orf12; HSPC014; HSPC036; hUMP1; mUMP1; PNAS-110; POMP; Proteasome maturation protein; proteassemblin; Protein UMP1 homolog; RGD1305831; UMP1; Voltage-gated K channel beta subunit 4.1; voltage-gated potassium channel beta subunit 4.1
Gene Symbols POMP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-141 of human POMP (NP_0570161).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51371
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.