Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POMT1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA566756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566756 Supplier Thermo Scientific Supplier No. PA566756

Rabbit Polyclonal Antibody

Immunogen sequence: GSTVWNVEEHR YGASQEQRER ERELHSPAQV DVSRNLSFMA RFSELQWRML ALRSDDSEHK YSSSPLEWVT LDTNIAYWLH PRTS Highest antigen sequence identity to the following orthologs - mouse 79%, rat 78%.

The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT2 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS) and limb-girdle muscular dystrophy type 2K (LGMD2K). Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POMT1
Gene Accession No. Q9Y6A1
Gene Alias AI505244; dolichyl-phosphate-mannose--protein mannosyltransferase 1; FLJ37239; LGMD2K; LOW QUALITY PROTEIN: protein O-mannosyl-transferase 1; MDDGA1; MDDGB1; MDDGC1; POMT1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1; RT; testis tissue sperm-binding protein Li 57p
Gene Symbols POMT1
Host Species Rabbit
Immunogen Recombinant Human POMT1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10585
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.