Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208627
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8627 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8627 Supplier Fabgennix Inc Supplier No. POMT1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POMT1 Polyclonal antibody specifically detects POMT1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen POMT1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene POMT1
Gene Accession No. Q8R2R1, Q99PR0, Q9Y6A1
Gene Alias AI505244; dolichyl-phosphate-mannose--protein mannosyltransferase 1; FLJ37239; LGMD2K; LOW QUALITY PROTEIN: protein O-mannosyl-transferase 1; MDDGA1; MDDGB1; MDDGC1; POMT1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1; RT; testis tissue sperm-binding protein Li 57p
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 400-460 on human POMT1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10585, 84430, 99011
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.