Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551987
Description
Immunogen sequence: GDGFFSSAFQ ARLSGNNLHN ASIPEHLAYG SVITVKNLRM AIGYLHSHRH LYPEGIGARQ QQVTTYLHKD YNNLWIIKKH NTNSDPLDPS FPVEFVRHGD IIRLEHKETS RNLHSHYHEA PMTRKHYQVT GYGINGT Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS).
Specifications
POMT2 | |
Polyclonal | |
Unconjugated | |
POMT2 | |
A830009D15Rik; AW046274; DKFZp686G10254; Dolichyl-phosphate-mannose--protein mannosyltransferase; dolichyl-phosphate-mannose--protein mannosyltransferase 2; FLJ22309; LGMD2N; MDDGA2; MDDGB2; MDDGC2; Pomt2; protein O-mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
217734, 29954, 688673 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BGQ4, Q9UKY4 | |
POMT2 | |
Recombinant protein corresponding to Human POMT2. Recombinant protein control fragment (Product #RP-88941). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction