Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMZP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531968 Supplier Invitrogen™ Supplier No. PA531968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene appears to have resulted from a fusion of DNA sequences derived from 2 distinct loci, specifically through the duplication of two internal exons from the POM121 gene and four 3' exons from the ZP3 gene. The 5' end of this gene is similar to the 5' coding region of the POM121 gene which encodes an integral nuclear pore membrane protein. However, the protein encoded by this gene lacks the nuclear pore localization motif. The 3' end of this gene is similar to the last 4 exons of the zona pellucida glycoprotein 3 (ZP3) gene and the encoded protein retains one zona pellucida domain. Multiple protein isoforms are encoded by transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMZP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene POMZP3
Gene Accession No. Q6PJE2
Gene Alias POM (POM121 homolog, rat) and ZP3 fusion; POM (POM121 rat homolog) and ZP3 fusion; POM121; POM121 and ZP3 fusion; POM121 and ZP3 fusion protein; POM121/ZP3 fusion protein; POMZP3; POM-ZP3; POM-ZP3 fusion protein
Gene Symbols POMZP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human POMZP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.