Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PON1 Antibody (4G8D3), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223610AF532
Description
PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PON1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PON1 | |
Partial recombinant human PON1 (between residues 1-200) expressed in E. coli [Uniprot P27169] | |
0.1 mL | |
Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
4G8D3 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction