Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PON1 Antibody (SR1678), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32176350UL
Description
PON1 Monoclonal antibody specifically detects PON1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PON1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1678 | |
Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1 | |
A synthesized peptide derived from human PON1 (Uniprot #: P27169) | |
50 μg | |
Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
5444 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction