Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PON1 Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502610

Description
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson′s disease.Specifications
PON1 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P27169 | |
PON1 | |
Full length human recombit protein of human PON1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2A6 | |
Unconjugated | |
PON1 | |
ESA, MVCD5, PON | |
Mouse | |
Affinity Chromatography | |
RUO | |
5444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction