Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PON1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519107
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519107 Supplier Invitrogen™ Supplier No. PA519107
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PON1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PON1
Gene Accession No. P27169
Gene Alias 2810004E20; 6330405I24Rik; A esterase 1; A-esterase 1; A-esterase 2; AI481612; AI786302; Aromatic esterase 1; aromatic esterase 2; arylesterase 1; arylesterase 2; arylesterase 3; arylesterase B-type; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; ESA; ESAesterase A; esterase A; K-45; MVCD5; paraoxanase-3; paraoxonase 1; paraoxonase 2; paraoxonase 3; paraoxonase B-type; paraoxonase nirs; PON; PON 1; PON 2; Pon1; PON2; PON3; serum aryldiakylphosphatase; serum aryldialkylphosphatase 1; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 1; Serum paraoxonase/arylesterase 2; serum paraoxonase/lactonase 3
Gene Symbols PON1
Host Species Goat
Immunogen Synthetic peptide sequence (EEPKVTQVYAEN) corresponding to the internal amino acids of PON1 (aa 313-325).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5444
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.