Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PON1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524178
Description
PON1 Polyclonal Antibody for Western Blot, IHC (P), Flow
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
Specifications
| PON1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P27169 | |
| PON1 | |
| KLH conjugated synthetic peptide between 118-145 amino acids from the central region of human PON1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PON1 | |
| 2810004E20; 6330405I24Rik; A esterase 1; A-esterase 1; A-esterase 2; AI481612; AI786302; Aromatic esterase 1; aromatic esterase 2; arylesterase 1; arylesterase 2; arylesterase 3; arylesterase B-type; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; ESA; ESAesterase A; esterase A; K-45; MVCD5; paraoxanase-3; paraoxonase 1; paraoxonase 2; paraoxonase 3; paraoxonase B-type; paraoxonase nirs; PON; PON 1; PON 2; Pon1; PON2; PON3; serum aryldiakylphosphatase; serum aryldialkylphosphatase 1; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 1; Serum paraoxonase/arylesterase 2; serum paraoxonase/lactonase 3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5444 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction