Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PON2 Recombinant Rabbit Monoclonal Antibody (JG35-80)
GREENER_CHOICE

Catalog No. PIMA534737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534737 Supplier Invitrogen™ Supplier No. MA534737
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: A549, human colon cancer tissue, human appendix tissue, human kidney tissue, mouse testis tissue.

This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PON2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone JG35-80
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene PON2
Gene Accession No. Q15165
Gene Alias 6330405I24Rik; A esterase 2; A-esterase 2; AI481612; aromatic esterase 2; arylesterase 2; paraoxonase 2; paraoxonase nirs; PON 2; PON2; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 2
Gene Symbols PON2
Host Species Rabbit
Immunogen Recombinant protein within C-terminal Human PON2 aa 160-354.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5445
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.