Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PON3 Antibody (OTI5C3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273530
Description
PON3 Monoclonal specifically detects PON3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PON3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, paraoxanase-3, paraoxonase 3, serum paraoxonase/lactonase 3 | |
Mouse | |
39.4 kDa | |
100 μg | |
5446 | |
Human | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI5C3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PON3 | |
Human recombinant protein fragment corresponding to amino acids 30-319 of human PON3(NP_000931) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction