Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PON3 Antibody (OTI5C3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273530H
Description
PON3 Monoclonal specifically detects PON3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PON3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PON3 | |
Recombinant protein fragment corresponding to amino acids 30-319 of human PON3 (NP_000931) produced in E.coli. | |
0.1 mL | |
5446.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5C3 | |
Western Blot, Immunohistochemistry | |
EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, paraoxanase-3, paraoxonase 3, serum paraoxonase/lactonase 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction