Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PON3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981664
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1664 Supplier Bioss Supplier No. BS11782R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Paraoxonase-1 (PON1) is a calcium-dependent ester hydrolase that is tightly associated with apoA-I in HDL. PON1 prevents the oxidation of LDL, thereby the formation of atherogenic oxidised-LDL. Polymorphisms of the human PON1 gene are correlated with coronary artery disease, indicating a genetic association between PON1 and coronary artery disease. PON1 activity is lower in subjects prone to development of atherosclerosis such as in type 1 and type 2 diabetes, familial hypercholesterolemia, and renal disease, indicating the involvement of PON1 in atherosclerosis development. PON2 is a ubiquitously expressed intracellular protein that can protect cells against oxidative damage. PON3 is similar to PON1 in activity but differs from it in substrate specificity. PON1 and PON3 are implicated in lowering the risk of developing coronary artery disease and atherosclerosis. Human PON3 protein shares the 3 conserved cysteine residues identified in PON1, suggesting their importance of in vivo activities. PON1-based catalytic scavengers can be a way to safe and effective organophosphate intoxication.

Specifications

Antigen PON3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PON3
Gene Accession No. Q15166
Gene Alias 2810004E20; AI786302; arylesterase 3; BOS_4015; paraoxanase-3; paraoxonase 3; PON3; Serum paraoxonase/lactonase 3
Gene Symbols PON3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PON3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5446
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.