Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ponceau 3R, Spectrum™ Chemical
SDP

Catalog No. 18612426 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-426 100 g Amber Glass Bottle
18-612-428 5 g Amber Glass Bottle
18-612-427 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-612-426 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO144100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3564-09-8

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 100 g
Chemical Name or Material Ponceau 3R
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.