Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Pondwater Tour Kit
SDPSCI_ED

Catalog No. p-2381090
Click to view available options
Type:
Pond Water Tour Kit
Reagent Refill

Includes

  • Complete lecture materials
  • Illustrated handouts
  • Teacher tips
  • Test procedures
  • TesTabs™ reagents
  • Data sheets
  • Wrap-up games

Study and measure changes in lake, stream, pond or aquarium water.

  • Helps teachers guide students through the study and measurement of changes in lake, stream, pond or aquarium water
  • Measures dissolved oxygen, pH, nitrate and ammonia

Order Info

Grades 4 to 8. Class size: 50 students.

Specifications

For Use With (Application) Study and measure changes in lake, stream, pond or aquarium water
For Use With (Equipment) Pond water tour kit
Includes Complete lecture materials, illustrated handouts, teacher tips, test procedures, testabs™ reagents, data sheets, wrap up games
Sufficient For 50 Students
Type Reagent Refill
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.