Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pontacyl Carmine 2B 93.0+%, TCI America™
SDP

Catalog No. P0595500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 6625-46-3
Molecular Formula C19H15N3Na2O9S2
Molecular Weight (g/mol) 539.441
MDL Number MFCD00059131
InChI Key JIAWPRMYSNTTFB-UHFFFAOYSA-L
PubChem CID 73555245
IUPAC Name disodium;5-acetamido-3-[(2-methoxyphenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES CC(=O)NC1=C2C(=CC(=C1)S(=O)(=O)[O-])C=C(C(=NNC3=CC=CC=C3OC)C2=O)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Red
Quantity 500 g
Formula Weight 539.44
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pontacyl Carmine 2B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.