Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers
Click to view available options
Quantity:
1 x 28 mL
5 x 28 mL
10 x 28 mL
30 x 28 mL
Description
Includes
Each bottle contains enough polymer for 960 samples
- A separation matrix for the 3730 and 3730xl DNA Analyzers
- Customized for DNA sequencing and fragment analysis.
- Pre-formulated to save time and reproducibility by ensuring consistent formulation
- Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
- Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
- Sufficient for 8,000 to 12,000 samples
Use With
- 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
- 3730 Running Buffer, 10X
- BigDye™ Terminator v1.1 Cycle Sequencing Kit
- BigDye™ Terminator v3.1 Cycle Sequencing Kit
- Hi-Di™ Formamide
Specifications
Specifications
| Accuracy | 98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary) |
| Content And Storage | Refrigerate at 2°C to 8°C. |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Standard Sequencing, Fragment Analysis (Denaturing) |
| On Instrument Life Usage | Stability on instrument 7 days, sufficient for 8000 to 12,000 samples |
| Product Line | POP-7™ |
| Product Type | Polymer |
| Quantity | 1 x 28 mL |
| Test Time | 1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction