Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Scientific Pop-Up Butterfly Cage
SDPSCI_ED

Catalog No. S02473
Click to view available options
:
Product Type: Net cage
This item is not returnable. View return policy
This item is not returnable. View return policy

This pop-up butterfly cage allows children, students, and butterfly enthusiasts to witness the wonder of the butterfly life cyle. The cage is useful as a tool for teaching about insects or life cycles.

  • Height: 24 in.
  • Diameter: 15 in.
  • Trim colors and pattern may vary
  • Child- and insect-friendly

Specifications

Color Variable
Product Type Net Cage
Height (English) 24 in.
Diameter (English) Outer 15 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.