Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988828
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8828 Supplier Bioss Supplier No. BS9107R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP.

Specifications

Antigen POP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pop1
Gene Accession No. Q99575
Gene Alias 4932434G09Rik; hPOP1; KIAA0061; POP1; POP1 homolog, ribonuclease P/MRP subunit; processing of precursor 1, ribonuclease P/MRP family, (S. cerevisiae); processing of precursor 1, ribonuclease P/MRP subunit; processing of precursor 1, ribonuclease P/MRP subunit (S. cerevisiae); processing of precursors 1; ribonucleases P/MRP protein subunit homolog; Ribonucleases P/MRP protein subunit POP1
Gene Symbols Pop1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASC2/POP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10940
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.