Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144918 Supplier Invitrogen™ Supplier No. PA5144918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVIYHALSQK EANDSDVQPS GAQRAEAFVR AFLKRSTPRM SPQAREDQLQ RKAVVLEYFT RHKRKE Antigen sequence identity: Mouse 65%; Rat 70%.

POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POP4
Gene Accession No. O95707
Gene Alias 1110023P21Rik; hPOP4; hypothetical protein LOC553787; POP4; POP4 (processing of precursor , S. cerevisiae) homolog; POP4 homolog, ribonuclease P/MRP subunit; POP4 protein; processing of precursor 4, ribonuclease P/MRP family, (S. cerevisiae); processing of precursor 4, ribonuclease P/MRP subunit; processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein subunit p29; RPP29; zgc:110597
Gene Symbols POP4
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 3-68. Recombinant protein control fragment (Product #RP-109757).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10775
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.