Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541823
Description
Peptide sequence: EDRLKVIPKL NCVFTVETDG FISYIYGSKF QLRSSERSAK KFKAKGTIDL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Yeast: 91%; Zebrafish: 92%.
POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.
Specifications
| POP4 | |
| Polyclonal | |
| Unconjugated | |
| POP4 | |
| 1110023P21Rik; hPOP4; hypothetical protein LOC553787; POP4; POP4 (processing of precursor , S. cerevisiae) homolog; POP4 homolog, ribonuclease P/MRP subunit; POP4 protein; processing of precursor 4, ribonuclease P/MRP family, (S. cerevisiae); processing of precursor 4, ribonuclease P/MRP subunit; processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein subunit p29; RPP29; zgc:110597 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10775 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95707 | |
| POP4 | |
| Synthetic peptide directed towards the C-terminal of human POP4 (aa 171-220). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction