Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110200
Description
Immunogen sequence: MKSVIYHALS QKEANDSDVQ PSGAQRAEAF VRAFLKRSTP RMSPQAREDQ LQRKAVVLEY FTRHKRKEKK KKAKGLSARQ RRELRLFDIK PEQQRYSLFL PLHELWKQYI RDLCSGLKPD TQPQMIQAKL LKADLHGAII SVTKSKCPSY VGITGILLQE TKHIFKIITK EDRLKVIPKL NCVFTVETDG FISYIYGSKF QLRSSERSAK KFKAKGTIDL.
POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.
Specifications
| POP4 | |
| Polyclonal | |
| Unconjugated | |
| POP4 | |
| 1110023P21Rik; hPOP4; hypothetical protein LOC553787; POP4; POP4 (processing of precursor , S. cerevisiae) homolog; POP4 homolog, ribonuclease P/MRP subunit; POP4 protein; processing of precursor 4, ribonuclease P/MRP family, (S. cerevisiae); processing of precursor 4, ribonuclease P/MRP subunit; processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein subunit p29; RPP29; zgc:110597 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10775, 292831, 66161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95707, Q5M882, Q9CR08 | |
| POP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human POP4 (NP_006618.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction