Learn More
POP4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305673AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.Specifications
| POP4 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O95707 | |
| POP4 | |
| Between 170 and 220 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| POP4 | |
| hPOP4, processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae), RPP29 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10775 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.