Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561549 Supplier Invitrogen™ Supplier No. PA561549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEFYQLVWSA LPFITYLENK GHRYPCFFNT LHVGGTIRTC QKFLIQYNRR QLLILLQNCT DEGEREAIQK SVTRSCLLEE EEES Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:11413139, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POP5
Gene Accession No. Q969H6
Gene Alias 1500019J17Rik; 2700077E03Rik; AD-008; AW049989; fc69d03; hPop5; HSPC004; POP5; POP5 homolog, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP family; processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae); processing of precursor 5, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein 3; ribonuclease P/MRP protein subunit POP5; Ribonuclease P/MRP protein subunit POP5-like protein; Rnasep3; rpP2; RPP20; wu:fc69d03; wu:fy24c05; x0003; zgc:110616
Gene Symbols POP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POP5. Recombinant protein control fragment (Product #RP-97818).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51367
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.