Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POP5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595789
Description
Immunogen sequence: SIGFAVRYLN AYTGIVLLRC RKEFYQLVWS ALPFITYLEN KGHRYPCFFN TLHVGGTIRT CQKFLIQYNR RQLLILLQNC TDEGEREAIQ KSVTRSCLLE E; Positive Samples: A-549, K562, 293T; Cellular Location: Nucleus, nucleolus.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:11413139, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
Specifications
POP5 | |
Polyclonal | |
Unconjugated | |
POP5 | |
1500019J17Rik; 2700077E03Rik; AD-008; AW049989; fc69d03; hPop5; HSPC004; POP5; POP5 homolog, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP family; processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae); processing of precursor 5, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein 3; ribonuclease P/MRP protein subunit POP5; Ribonuclease P/MRP protein subunit POP5-like protein; Rnasep3; rpP2; RPP20; wu:fc69d03; wu:fy24c05; x0003; zgc:110616 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117241, 51367 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q969H6 | |
POP5 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human POP5 (NP_0570022). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction