Learn More
POP5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305746AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP.Specifications
POP5 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q969H6 | |
POP5 | |
Between 113 and 163 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
POP5 | |
hPop5, HSPC004, processing of precursor 5, ribonuclease P/MRP subunit, processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae), RPP2, RPP20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51367 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.