Learn More
Invitrogen™ POP7 Monoclonal Antibody (3F12)

Description
Peptide Sequence: MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINH.
Specifications
Specifications
| Antigen | POP7 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3F12 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | POP7 |
| Gene Accession No. | O75817 |
| Gene Alias | 0610037N12Rik; hPOP7; POP7; POP7 (processing of precursor, S. cerevisiae) homolog; POP7 homolog, ribonuclease P/MRP subunit; processing of precursor 7, ribonuclease P subunit; processing of precursor 7, ribonuclease P/MRP subunit; ribonuclease P protein subunit p20; Ribonucleases P/MRP protein subunit POP7 homolog; RNaseP protein p20; RPP2; RPP20 |
| Gene Symbols | POP7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.