Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POPDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555029 Supplier Invitrogen™ Supplier No. PA555029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TAETSCSYIS WPRKSLHLLL TKERYISCLF SALLGYDISE KLYTLNDKLF AKFGLRFDIR LPSLYHVLGP TAADAGPESE KGDEEVCEPA VSPPQATPTS LQQTPPCSTP PATTNFPAPP T Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

This gene encodes a member of the POP family of proteins which contain three putative transmembrane domains. This membrane associated protein is predominantly expressed in skeletal and cardiac muscle, and may have an important function in these tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POPDC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POPDC2
Gene Accession No. Q9HBU9
Gene Alias AV006127; Pop2; POPDC2; popeye 2; popeye domain containing 2; popeye domain-containing 2; Popeye domain-containing protein 2; popeye protein 2
Gene Symbols POPDC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POPDC2. Recombinant protein control fragment (Product #RP-90944).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.