Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POPDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543406 Supplier Invitrogen™ Supplier No. PA543406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YLLFAQHRYI SRLFSVLIGS DIADKLYALN DRVYIGKRYH YDIRLPNFYQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

POPDC3 is a member of the POP family of proteins containing three putative transmembrane domains. The protein is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POPDC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POPDC3
Gene Accession No. Q9HBV1
Gene Alias AA682104; bA355M14.1; MGC22671; POP3; Popdc3; popeye 3; popeye domain containing 3; popeye domain-containing 3; popeye domain-containing protein 3; Popeye protein 3; RP11-99L11.2
Gene Symbols POPDC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human POPDC3 (aa 218-267).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64208
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.