Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POPDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115782 Supplier Invitrogen™ Supplier No. PA5115782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total POPDC3.

POPDC3 is a member of the POP family of proteins containing three putative transmembrane domains. The protein is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POPDC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene POPDC3
Gene Accession No. Q9ES81, Q9HBV1
Gene Alias AA682104; bA355M14.1; MGC22671; POP3; Popdc3; popeye 3; popeye domain containing 3; popeye domain-containing 3; popeye domain-containing protein 3; Popeye protein 3; RP11-99L11.2
Gene Symbols POPDC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human POPDC3(Accession Q9HBV1), corresponding to amino acid residues Y258-K291.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641520, 64208, 78977
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.