Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POR/Cytochrome P450 Reductase Antibody (OTI3F10), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273539PCP
Description
POR/Cytochrome P450 Reductase Monoclonal specifically detects POR/Cytochrome P450 Reductase in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| POR/Cytochrome P450 Reductase | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| POR | |
| Full length recombinant protein of human POR/Cytochrome P450 Reductase (NP_000932) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI3F10 | |
| Flow Cytometry | |
| CPR, CYPORFLJ26468, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Stem Cell Markers | |
| 5447.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction