Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Porcelain Spatula with Spoon
SDPSCI_ED

Catalog No. S139693 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
0.1 mL
0.3 mL
0.5 mL
1.2 mL
5.5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139693 0.1 mL
S139694 0.3 mL
S139695 0.5 mL
S139696 1.2 mL
S139697 5.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S139693 Supplier United Scientific™ Supplier No. JSS001
Add to Cart
Add to Cart
This item is not returnable. View return policy

These spatulas offer laboratory grade porcelainware at substantial cost savings.

  • Glazed porcelain laboratory spatula designed with one flat end and one spoon end
  • Available in five different sizes

Specifications

Quantity 1 Spatula
Shape Flat Squared End, Spoon End
Autoclavable Autoclavable
Capacity (Metric) 0.1 mL
Product Type Spatula
Material Porcelain
Length (Metric) 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.