Learn More
United Scientific™ Porcelainware Starter Kit
SDPSCI_ED
Catalog No. S139691 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Porcelainware Starter Kit; Autoclavable: Autoclavable
Catalog No.
S139691 Porcelainware Starter Kit; Autoclavable: Autoclavable
1 options
This item is not returnable. View return policy
Catalog No. S139691 Supplier United Scientific™ Supplier No. PORTKIT2
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Porcelainware Starter Kit includes everything you need for a lab station requiring porcelain labware.
Convenient collection of porcelain products and accessories commonly used in the laboratory.
  • Contains one each of the following items:
    • 50 mL low-form ceramic crucible (JCL050)
    • 50 mL high-form ceramic crucible (JCT050)
    • 200 mL ceramic casserole (JPC200)
    • 275 mL ceramic mortar and pestle (JMD275)
    • 80 mL ceramic evaporating dish (JED080)
    • 2 in. clay pipe triangle (CPT020)
    • Nickel-plated steel crucible tongs, 9 in.(CTSP09)
Specifications
Material Porcelain
Capacity (Metric) Various
Autoclavable Autoclavable
Diameter (Metric) Outer Various
Height (Metric) Various
Product Type Porcelainware Kit
Shape Assorted
Diameter (Metric) Base Various