Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine CD34 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $573.00
Specifications
Antigen | CD34 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CD34 Polyclonal specifically detects CD34 in Porcine samples. It is validated for Western Blot.Specifications
CD34 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Pig | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
CD34 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_999251 | |
947 | |
Mouse myeloma cell line NS0-derived recombinant porcine CD34 Ala20-Thr294 (Thr32Ala, Ala52Thr, Pro71Ser, Val126Ala, His278Asn) Accession # NP_999251 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects porcine CD34 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant rat CD34 is observed, and less than 5% cross-reactivity with recombinant mouse CD34 and recombinant human CD34 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title