Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine Circovirus Type 2 Capsid Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534968 Supplier Invitrogen™ Supplier No. PA534968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Porcine circovirus type 2 Capsid-transfected Sf21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The PCV2 virion measures approximately 17 nm in diameter, is non-enveloped and consists of a circular single-stranded DNA surrounded by an icosahedral capsid. The ambisense DNA molecule contains about 1.77 kb and 11 putative open reading frames (ORFs). PCV2 has been associated with a number of disease syndromes in pigs. Post-weaning multisystemic wasting syndrome (PMWS) is an important emerging viral disease affecting nursery and grower pigs, which has recently been associated with porcine circovirus type 2 (PCV2) infections. PMWS has been reported in Europe, South East Asia, and the Americas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Porcine Circovirus Type 2 Capsid
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias circovirus 2; Circovirus capsid; PCV2; PCV2 capsid; Porcine Circovirus
Host Species Rabbit
Immunogen Recombinant fragment of Porcine circovirus type 2 Capsid protein (Strain: swPCV2bps1).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.